MilliporeSigma
  • Micronization increases the bioaccessibility of polyphenols from granulometrically separated olive pomace fractions.
Micronization increases the bioaccessibility of polyphenols from granulometrically separated olive pomace fractions.
Food chemistry (2020-12-06)
Caroline Sefrin Speroni, Daniela Rigo Guerra, Ana Betine Beutinger Bender, Jessica Stiebe, Cristiano Augusto Ballus, Leila Picolli da Silva, Jesús Lozano-Sánchez, Tatiana Emanuelli
ABSTRACT
The effect of micronization of granulometrically fractionated olive pomace (OP) on the bioaccessibility of polyphenols and the antioxidant capacity was investigated during sequential in vitro static digestion. Crude OP was fractionated in a 2-mm sieve (F1: > 2 mm; F2: < 2 mm) and then micronized (300 r min-1, 5 h) generating F1AG (17.8 μm) and F2AG (15.6 μm). Micronization increased the release of hydroxytyrosol, oleuropein, caffeic acid, and decarboxymethyl oleuropein aglycone (3,4-DHPEA-EDA) in the salivary and gastric phase, beyond luteolin in the gastric phase. Micronization also increased the intestinal bioaccessibility of hydroxytyrosol, 3,4-DHPEA-EDA, oleuropein, luteolin, and apigenin; it was more effective for F2AG than F1AG. Micronized samples increased antioxidant capacity in the gastric phase. F2AG exhibited the highest antioxidant capacity in the insoluble intestinal fraction. Thus, micronization can be further exploited to improve the nutraceutical properties of OP by increasing the bioaccessibility and antioxidant capacity of phenolic compounds.
Product Description
Millipore
Bile salts, suitable for microbiology
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, ≥250 units/mg solid
Sigma-Aldrich
α-Amylase from porcine pancreas, PMSF Treated, Type I-A, saline suspension, ≥1000 units/mg protein (E1%/280)
Sigma-Aldrich
Pancreatin from porcine pancreas, 8 × USP specifications
Sigma-Aldrich
(±)-6-Hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid, 97%