MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Synthesis and evaluation of anti-tubercular activity of new dithiocarbamate sugar derivatives.
Synthesis and evaluation of anti-tubercular activity of new dithiocarbamate sugar derivatives.
Bioorganic & medicinal chemistry letters (2011-01-15)
Yasuhiro Horita, Takemasa Takii, Ryuji Kuroishi, Taku Chiba, Kenji Ogawa, Laurent Kremer, Yasuo Sato, YooSa Lee, Tomohiro Hasegawa, Kikuo Onozaki
ABSTRACT
The present study was undertaken to optimize the anti-tubercular activity of 2-acetamido-2-deoxy-β-D-glucopyranosyl N,N-dimethyldithiocarbamate (OCT313, Glc-NAc-DMDC), a lead compound previously reported by us. Structural modifications of OCT313 included the replacements of the DMDC group at C-1 by pyrrolidine dithiocarbamate (PDTC) and the acetyl group at C-2 by either propyl, butyl, benzyl or oleic acid groups. The antimycobacterial activities of these derivatives were evaluated against Mycobacterium tuberculosis (MTB). Glc-NAc-pyrrolidine dithiocarbamate (OCT313HK, Glc-NAc-PDTC) exhibited the most potent anti-tubercular activity with the minimal inhibitory concentration (MIC) of 6.25-12.5 μg/ml. The antibacterial activity of OCT313HK was highly specific to MTB and Mycobacterium bovis BCG, but not against Mycobacterium avium, Mycobacterium smegmatis, Staphylococcus aureus or Escherichia coli. Importantly, OCT313HK was also effective against MTB clinical isolates, including multidrug-resistant (MDR) and extensively drug-resistant (XDR) strains. Interestingly, OCT313HK was exerted the primary bactericidal activity, and it was also exhibited the bacteriolytic activity at high concentrations. We next investigated whether the mycobacterial monooxygenase EthA, a common activator of thiocarbamide-containing anti-tubercular drugs, also activated OCT313HK. Contrary to our expectations, the anti-tubercular activity of dithiocarbamate sugar derivatives and dithiocarbamates were not dependent on ethA expression, in contrast to thiocarbamide-containing drugs. Overall, this study presents OCT313HK as a novel and potent compound against MTB, particularly promising to overcome drug resistance.
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
Ammonium pyrrolidinedithiocarbamate, ~99%
Supelco
Isoniazid, analytical standard, ≥99% (TLC)
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)