MilliporeSigma
  • Synthesis and biological evaluation of estradiol linked pyrrolo[2,1-c][1,4]benzodiazepine (PBD) conjugates as potential anticancer agents.
Synthesis and biological evaluation of estradiol linked pyrrolo[2,1-c][1,4]benzodiazepine (PBD) conjugates as potential anticancer agents.
Bioorganic & medicinal chemistry (2011-04-05)
Ahmed Kamal, M Kashi Reddy, M Janaki Ramaiah, Rajender, J Surendranadha Reddy, Y V V Srikanth, D Dastagiri, E Vijaya Bharathi, S N C V L Pushpavalli, Pranjal Sarma, Manika Pal-Bhadra
ABSTRACT
A series of new estradiol linked pyrrolo[2,1-c][1,4]benzodiazepine (E(2)-PBD) conjugates (3a-f, 4a-f and 5a-f) with different linker architectures including a triazole moiety have been designed and synthesized. All the 18 compounds have been evaluated for their anticancer activity and it is observed that some of the compounds particularly 4c-e and 5c,d exhibited significant anticancer activity. The detailed biological aspects relating to the cell cycle effects and tubulin depolymerization activity have been examined with a view to understand the mechanism of action of these conjugates. Among all these conjugates, one of the compound 5c could be considered as the most effective compound particularly against MCF-7 breast cancer cell line.
Sigma-Aldrich
Estradiol, meets USP testing specifications
Sigma-Aldrich
β-Estradiol, ≥98%
Sigma-Aldrich
β-Estradiol, analytical standard
Sigma-Aldrich
β-Estradiol, BioReagent, powder, suitable for cell culture
Sigma-Aldrich
β-Estradiol, powder, γ-irradiated, suitable for cell culture
Sigma-Aldrich
Doxorubicin hydrochloride, suitable for fluorescence, 98.0-102.0% (HPLC)
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)