MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Vasorelaxant effect of flavonoids through calmodulin inhibition: Ex vivo, in vitro, and in silico approaches.
Vasorelaxant effect of flavonoids through calmodulin inhibition: Ex vivo, in vitro, and in silico approaches.
Bioorganic & medicinal chemistry (2010-12-07)
Mariana Torres-Piedra, Mario Figueroa, Oswaldo Hernández-Abreu, Maximiliano Ibarra-Barajas, Gabriel Navarrete-Vázquez, Samuel Estrada-Soto
ABSTRACT
In our search for potential antihypertensive agents, a series of structurally-related flavonoids was screened. Ex vivo and in vitro biological evaluations indicated that compounds 1-7 displayed an important vasorelaxant effect on the endothelium-intact (E(+)) and -denuded (E(-)) aortic rings test. Their in vitro anti-calmodulin (CaM) properties were determined by means of the inhibitory effect on the activation of the calmodulin-sensitive cAMP phosphodiesterase (PDE1) assay. Molecular modeling experiments were also performed in order to explore the probable binding site of 1-7 with CaM, and the results indicated that they could bind to the protein in the same pocket as trifluoperazine (TFP), a well-known CaM inhibitor.
Sigma-Aldrich
Chrysin, ≥96.5%
Sigma-Aldrich
Nifedipine, ≥98% (HPLC), powder
Sigma-Aldrich
Carbamoylcholine chloride, ≥98% (titration), crystalline
Sigma-Aldrich
Flavone, ≥99.0%