MilliporeSigma
  • Simultaneous Targeting of RSK and AKT Efficiently Inhibits YB-1-Mediated Repair of Ionizing Radiation-Induced DNA Double-Strand Breaks in Breast Cancer Cells.
Simultaneous Targeting of RSK and AKT Efficiently Inhibits YB-1-Mediated Repair of Ionizing Radiation-Induced DNA Double-Strand Breaks in Breast Cancer Cells.
International journal of radiation oncology, biology, physics (2020-09-16)
Konstanze Lettau, Daniel Zips, Mahmoud Toulany
ABSTRACT
Y-box binding protein 1 (YB-1) overexpression is associated with chemotherapy- and radiation therapy resistance. Ionizing radiation (IR), receptor tyrosine kinase ligands, and mutation in KRAS gene stimulate activation of YB-1. YB-1 accelerates the repair of IR-induced DNA double-strand breaks (DSBs). Ribosomal S6 kinase (RSK) is the main kinase inducing YB-1 phosphorylation. We investigated the impact of RSK targeting on DSB repair and radiosensitivity. The triple negative breast cancer (TNBC) cell lines MDA-MB-231, MDA-MB-468, and Hs 578T, in addition to non-TNBC cell lines MCF7, HBL-100, and SKBR3, were used. MCF-10A cells were included as normal breast epithelial cells. The RSK inhibitor LJI308 was used to investigate the role of RSK activity in S102 phosphorylation of YB-1 and YB-1-associated signaling pathways. The activation status of the underlying pathways was investigated by Western blotting after treatment with pharmacologic inhibitors or transfection with siRNA. The impact of LJI308 on DSB repair and postirradiation cell survival was tested by the γH2AX foci and the standard clonogenic assays, respectively. LJI308 inhibited the phosphorylation of RSK (T359/S363) and YB-1 (S102) after irradiation, treatment with EGF, and in cells expressing a KRAS mutation. LJI308 treatment slightly inhibited DSB repair only in some of the cell lines tested. This was shown to be due to PI3K-dependent stimulation of AKT or constitutive AKT activity mainly in cancer cells but not in normal breast epithelial MCF-10A cells. Simultaneous targeting of AKT and RSK strongly blocked DSB repair in all cancer cell lines, independent of TNBC status or KRAS mutation, with a minor effect in MCF-10A cells. Cotargeting of RSK- and AKT-induced radiation sensitivity in TNBC MDA-MB-231 and non-TNBC MCF7 cells but not in MCF-10A cells. Simultaneous targeting of RSK and AKT might be an efficient approach to block the repair of DSBs after irradiation and to induce radiosensitization of breast cancer cells.
Sigma-Aldrich
MISSION® esiRNA, targeting human YBX1