MilliporeSigma
  • FUBP1 promotes colorectal cancer stemness and metastasis via DVL1-mediated activation of Wnt/β-catenin signaling.
FUBP1 promotes colorectal cancer stemness and metastasis via DVL1-mediated activation of Wnt/β-catenin signaling.
Molecular oncology (2021-07-22)
Haofan Yin, Tianxiao Gao, Jinye Xie, Zhijian Huang, Xiaoyan Zhang, Fengyu Yang, Weiwei Qi, Zhonghan Yang, Ti Zhou, Guoquan Gao, Xia Yang
ABSTRACT
Distant metastasis is, unfortunately, the leading cause of death in colorectal cancer (CRC). Approximately 50% of CRC patients develop liver metastases, while 10-30% of patients develop pulmonary metastases. The occurrence of metastasis is considered to be almost exclusively driven by cancer stem cells (CSCs) formation. However, the key molecules that confer the transformation to stem cells in CRC, and subsequent metastasis, remain unclear. Far upstream element-binding protein 1 (FUBP1), a transcriptional regulator of c-Myc, was screened in CSCs of CRC by mass spectrometry and was examined by immunohistochemistry in a cohort of CRC tissues. FUBP1 was upregulated in 85% of KRAS-mutant and 25% of wild-type CRC patients. Further, whether in KRAS-mutant or wild-type patients, elevated FUBP1 was positively correlated with CRC lymph node metastasis and clinical stage, and negatively associated with overall survival. Overexpression of FUBP1 significantly enhanced CRC cell migration, invasion, tumor sphere formation, and CD133 and ALDH1 expression in vitro, and tumorigenicity in vivo. Mechanistically, FUBP1 promoted the initiation of CSCs by activating Wnt/β-catenin signaling via directly binding to the promoter of DVL1, a potent activator of β-catenin. Knockdown of DVL1 significantly inhibited the transformation to stem cells in, as well as the tumorigenicity of, CRC. Activation of Wnt/β-catenin signaling by DVL1 increased pluripotent transcription factors, including c-Myc, NANOG, and SOX2. Moreover, FUBP1 was upregulated at the post-transcriptional level. Elevated FUBP1 levels in KRAS wild-type CRC patients is due to the decrease in Smurf2, which promotes ubiquitin-mediated degradation of FUBP1. In contrast, FUBP1 was upregulated in KRAS-mutant patients through both inhibition of caspase 3-dependent cleavage and decreased Smurf2. Our results demonstrate, for the first time, that FUBP1 is an oncogene, initiating the development of CSCs, as well as a new powerful endogenous Wnt-signaling agonist that could provide an important prognostic factor and therapeutic target for metastasis in both KRAS-mutant and wild-type CRC.
Sigma-Aldrich
Goat Anti-Mouse IgG Antibody, (H+L) HRP conjugate, 1 mg/mL, Chemicon®
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-FUSE-binding protein 1 Antibody, from rabbit, purified by affinity chromatography