MilliporeSigma
  • Repeated cocaine administration decreases 5-HT(2A) receptor-mediated serotonergic enhancement of synaptic activity in rat medial prefrontal cortex.
Repeated cocaine administration decreases 5-HT(2A) receptor-mediated serotonergic enhancement of synaptic activity in rat medial prefrontal cortex.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology (2009-02-13)
Chiung-Chun Huang, Ying-Ching Liang, Cheng-Che Lee, Mei-Ying Wu, Kuei-Sen Hsu
ABSTRACT
Neural adaptations in the medial prefrontal cortex (mPFC) are thought to be crucial in the development and maintenance of addictive behaviors. The mPFC receives a dense serotonergic (5-hydroxytryptamine, 5-HT) innervation from raphe nuclei and 5-HT exerts complex actions on mPFC pyramidal neurons. The present study, using a rat model of behavioral sensitization to cocaine, was designed to determine whether repeated cocaine exposure in vivo is capable of altering 5-HT-induced regulation of glutamatergic transmission in the mPFC. In layer V pyramidal neurons of the mPFC, application of 5-HT, through activation of 5-HT(2A) receptors, induced a massive enhancement of spontaneous excitatory postsynaptic currents (sEPSCs). Repeated cocaine administration for 5 days resulted in an attenuation in the ability of 5-HT to enhance sEPSCs. This effect was prevented when cocaine was co-administered with the selective 5-HT(2A) receptor antagonist ketanserin and was mimicked by repeated 5-HT(2A) receptor agonist (-)4-iodo-2,5-dimethoxyphenylisopropylamine administration. Repeated cocaine administration is not associated with any changes in the levels of 5-HT(2A) receptors or regulator of GTP-binding protein signaling 4. These results suggest that cocaine-induced inhibition of 5-HT(2A) receptor-mediated enhancement of glutamatergic transmission in the mPFC may be caused, at least in part, by the impairment of coupling of 5-HT(2A) receptors with GTP-binding proteins during cocaine withdrawal. These alterations in 5-HT(2A) receptor responsiveness in the mPFC may be relevant to the development of behavioral sensitization and withdrawal effects following repeated cocaine administration.
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
1(S),9(R)-(−)-Bicuculline methiodide, ≥95.0% (HPCE)