MilliporeSigma
  • The molecular and metabolic program by which white adipocytes adapt to cool physiologic temperatures.
The molecular and metabolic program by which white adipocytes adapt to cool physiologic temperatures.
PLoS biology (2021-05-13)
Hiroyuki Mori, Colleen E Dugan, Akira Nishii, Ameena Benchamana, Ziru Li, Thomas S Cadenhead, Arun K Das, Charles R Evans, Katherine A Overmyer, Steven M Romanelli, Sydney K Peterson, Devika P Bagchi, Callie A Corsa, Julie Hardij, Brian S Learman, Mahmoud El Azzouny, Joshua J Coon, Ken Inoki, Ormond A MacDougald
ABSTRACT
Although visceral adipocytes located within the body's central core are maintained at approximately 37°C, adipocytes within bone marrow, subcutaneous, and dermal depots are found primarily within the peripheral shell and generally exist at cooler temperatures. Responses of brown and beige/brite adipocytes to cold stress are well studied; however, comparatively little is known about mechanisms by which white adipocytes adapt to temperatures below 37°C. Here, we report that adaptation of cultured adipocytes to 31°C, the temperature at which distal marrow adipose tissues and subcutaneous adipose tissues often reside, increases anabolic and catabolic lipid metabolism, and elevates oxygen consumption. Cool adipocytes rely less on glucose and more on pyruvate, glutamine, and, especially, fatty acids as energy sources. Exposure of cultured adipocytes and gluteal white adipose tissue (WAT) to cool temperatures activates a shared program of gene expression. Cool temperatures induce stearoyl-CoA desaturase-1 (SCD1) expression and monounsaturated lipid levels in cultured adipocytes and distal bone marrow adipose tissues (BMATs), and SCD1 activity is required for acquisition of maximal oxygen consumption at 31°C.
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium, Without glucose, L-glutamine, phenol red, sodium pyruvate and sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
Anti-Adiponectin antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Free Glycerol Determination Kit, 1 kit sufficient for 1000 reactions
Sigma-Aldrich
Anti-PPAR γ Antibody, isoform 1&2, ascites fluid, clone 1H4, Chemicon®