MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Efficacy and long-term safety of CRISPR/Cas9 genome editing in the SOD1-linked mouse models of ALS.
Efficacy and long-term safety of CRISPR/Cas9 genome editing in the SOD1-linked mouse models of ALS.
Communications biology (2021-03-27)
Han-Xiang Deng, Hong Zhai, Yong Shi, Guoxiang Liu, Jessica Lowry, Bin Liu, Éanna B Ryan, Jianhua Yan, Yi Yang, Nigel Zhang, Zhihua Yang, Erdong Liu, Yongchao C Ma, Teepu Siddique
ABSTRACT
CRISPR/Cas9-mediated genome editing provides potential for therapeutic development. Efficacy and long-term safety represent major concerns that remain to be adequately addressed in preclinical studies. Here we show that CRISPR/Cas9-mediated genome editing in two distinct SOD1-amyotrophic lateral sclerosis (ALS) transgenic mouse models prevented the development of ALS-like disease and pathology. The disease-linked transgene was effectively edited, with rare off-target editing events. We observed frequent large DNA deletions, ranging from a few hundred to several thousand base pairs. We determined that these large deletions were mediated by proximate identical sequences in Alu elements. No evidence of other diseases was observed beyond 2 years of age in these genome edited mice. Our data provide preclinical evidence of the efficacy and long-term safety of the CRISPR/Cas9 therapeutic approach. Moreover, the molecular mechanism of proximate identical sequences-mediated recombination provides mechanistic information to optimize therapeutic targeting design, and to avoid or minimize unintended and potentially deleterious recombination events.
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Actin (20-33) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Choline Acetyltransferase Antibody, Chemicon®, from goat
Sigma-Aldrich
Anti-Iba1/AIF1 Antibody, clone 20A12.1, from mouse