MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Spatiotemporally confined red light-controlled gene delivery at single-cell resolution using adeno-associated viral vectors.
Spatiotemporally confined red light-controlled gene delivery at single-cell resolution using adeno-associated viral vectors.
Science advances (2021-06-18)
Maximilian Hörner, Carolina Jerez-Longres, Anna Hudek, Sebastian Hook, O Sascha Yousefi, Wolfgang W A Schamel, Cindy Hörner, Matias D Zurbriggen, Haifeng Ye, Hanna J Wagner, Wilfried Weber
ABSTRACT
Methodologies for the controlled delivery of genetic information into target cells are of utmost importance for genetic engineering in both fundamental and applied research. However, available methods for efficient gene transfer into user-selected or even single cells suffer from low throughput, the need for complicated equipment, high invasiveness, or side effects by off-target viral uptake. Here, we engineer an adeno-associated viral (AAV) vector system that transfers genetic information into native target cells upon illumination with cell-compatible red light. This OptoAAV system allows adjustable and spatially resolved gene transfer down to single-cell resolution and is compatible with different cell lines and primary cells. Moreover, the sequential application of multiple OptoAAVs enables spatially resolved transduction with different transgenes. The approach presented is likely extendable to other classes of viral vectors and is expected to foster advances in basic and applied genetic research.
Sigma-Aldrich
McCoy′s 5A Medium, Modified, with sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
hEGF, EGF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Roche
cOmplete Protease Inhibitor Cocktail, Tablets provided in EASYpacks