MilliporeSigma
  • Drosophila as a useful model for understanding the evolutionary physiology of obesity resistance and metabolic thrift.
Drosophila as a useful model for understanding the evolutionary physiology of obesity resistance and metabolic thrift.
Fly (2021-03-12)
Lindsey J Gray, Marla B Sokolowski, Stephen J Simpson
ABSTRACT
Evolved metabolic thriftiness in humans is a proposed contributor to the obesity epidemic. Insect models have been shown to evolve both 'metabolic thrift' in response to rearing on high-protein diets that promote leanness, and 'obesity resistance' when reared on fattening high-carbohydrate, low-protein foods. Despite the hypothesis that human obesity is caused by evolved metabolic thrift, genetic contributions to this physiological trait remain elusive. Here we conducted a pilot study to determine whether thrift and obesity resistance can arise under laboratory based 'quasi-natural selection' in the genetic model organism Drosophila melanogaster. We found that both these traits can evolve within 16 generations. Contrary to predictions from the 'thrifty genotype/phenotype' hypothesis, we found that when animals from a metabolic thrift inducing high-protein environment are mismatched to fattening high-carbohydrate foods, they did not become 'obese'. Rather, they accumulate less triglyceride than control animals, not more. We speculate that this may arise through as yet un-quantified parental effects - potentially epigenetic. This study establishes that D. melanogaster could be a useful model for elucidating the role of the trans- and inter-generational effects of diet on the genetics of metabolic traits in higher animals.
Sigma-Aldrich
Casein from bovine milk, technical grade