MilliporeSigma
  • Weight loss and cystic disease progression in autosomal dominant polycystic kidney disease.
Weight loss and cystic disease progression in autosomal dominant polycystic kidney disease.
iScience (2022-01-22)
Katharina Hopp, Victoria A Catenacci, Nidhi Dwivedi, Timothy L Kline, Wei Wang, Zhiying You, Dustin T Nguyen, Kristen Bing, Bhavya Poudyal, Ginger C Johnson, Matthew R Jackman, Marsha Miller, Cortney N Steele, Natalie J Serkova, Paul S MacLean, Raphael A Nemenoff, Berenice Gitomer, Michel Chonchol, Kristen L Nowak
ABSTRACT
Progression of autosomal dominant polycystic kidney disease (ADPKD) is modified by metabolic defects and obesity. Indeed, reduced food intake slows cyst growth in preclinical rodent studies. Here, we demonstrate the feasibility of daily caloric restriction (DCR) and intermittent fasting (IMF) in a cohort of overweight or obese patients with ADPKD. Clinically significant weight loss occurred with both DCR and IMF; however, weight loss was greater and adherence and tolerability were better with DCR. Further, slowed kidney growth correlated with body weight and visceral adiposity loss independent of dietary regimen. Similarly, we compared the therapeutic efficacy of DCR, IMF, and time restricted feeding (TRF) using an orthologous ADPKD mouse model. Only ADPKD animals on DCR lost significant weight and showed slowed cyst growth compared to ad libitum, IMF, or TRF feeding. Collectively, this supports therapeutic feasibility of caloric restriction in ADPKD, with potential efficacy benefits driven by weight loss.
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution