MilliporeSigma
  • Increased nitric oxide (NO) production by antigen-presenting dendritic cells is responsible for low allogeneic mixed leucocyte reaction (MLR) in primary biliary cirrhosis (PBC).
Increased nitric oxide (NO) production by antigen-presenting dendritic cells is responsible for low allogeneic mixed leucocyte reaction (MLR) in primary biliary cirrhosis (PBC).
Clinical and experimental immunology (1998-10-09)
K Yamamoto, S M Akbar, T Masumoto, M Onji
ABSTRACT
The levels of blastogenesis in allogeneic MLR containing T cells from one normal volunteer and irradiated dendritic cells from 29 patients with PBC, 17 patients with chronic hepatitis type C (CH-C) and 22 allogeneic normal controls were compared to see if there is any role of antigen-presenting cells (APC) in the pathogenesis of PBC. The stimulatory capacity of dendritic cells from PBC was significantly lower compared with that of dendritic cells from CH-C (P < 0.05) and normal controls (P < 0.05), which could not be attributable either to the levels of expression of surface molecules, such as HLA-DR and CD86 on dendritic cells, or to the levels of cytokines, such as IL-10 and IL-12. Significantly higher levels of NO were seen in the allogeneic MLR supernatants containing dendritic cells from PBC compared with the supernatants from cultures containing dendritic cells from CH-C (P < 0.001) or normal controls (P < 0.001). Moreover, dendritic cells from PBC produced 10 times more NO compared with dendritic cells from CH-C and normal controls (21.9 +/- 2.8 microM versus 1.6 +/- 0.3 microM and 1.6 +/- 0.3 microM, respectively; P < 0.001). The addition of N(G)-monomethyl-L-arginine monoacetate (L-NMMA), a known inhibitor of NO in allogeneic MLR containing dendritic cells from PBC, resulted in a significant decrease of NO and increase of blastogenesis. The selective impairment of dendritic cell function, increased production of NO by dendritic cells and restoration of blastogenesis using NO inhibitor in PBC have suggested a role for NO and dysfunction of dendritic cells in the pathogenesis of PBC. This inspires optimism that modulating the function of dendritic cells and controlling NO production, an improved therapeutic approach, might be planned for PBC.
Sigma-Aldrich
PANSORBIN® Cells, Standardized