MilliporeSigma
  • Microglia-Derived Olfactomedin-like 3 Promotes Pro-Tumorigenic Microglial Function and Malignant Features of Glioma Cells.
Microglia-Derived Olfactomedin-like 3 Promotes Pro-Tumorigenic Microglial Function and Malignant Features of Glioma Cells.
International journal of molecular sciences (2021-12-11)
Ryan G Toedebusch, Christopher A Lucchesi, Eshetu T Debebe, Luke A Wittenburg, Xinbin Chen, Christine M Toedebusch
ABSTRACT
Under the influence of transforming growth factor-beta (TGFβ), glioma-associated microglia produce molecules that promote glioma growth and invasion. Olfactomedin-like 3 (Olfml3), a novel, secreted glycoprotein, is known to promote several non-CNS cancers. While it is a direct TGFβ1 target gene in microglia, the role of microglia-derived OLFML3 in glioma progression is unknown. Here, we tested the hypotheses that microglial Olfml3 is integral to the pro-tumorigenic glioma-associated microglia phenotype and promotes glioma cell malignancy. Using an Olfml3 knockout microglial cell line (N9), we demonstrated that Olfml3 is a direct target gene of all TGFβ isoforms in murine microglia. Moreover, loss of Olfml3 attenuated TGFβ-induced restraint on microglial immune function and production of cytokines that are critical in promoting glioma cell malignancy. Importantly, microglia-derived OLFML3 directly contributes to glioma cell malignancy through increased migration and invasion. While exposure to conditioned medium (CM) from isogenic control microglia pre-treated with TGFβ increased mouse glioma cell (GL261) migration and invasion, this effect was abolished with exposure to CM from TGFβ-treated Olfml3-/- microglia. Taken together, our data suggest that Olfml3 may serve as a gatekeeper for TGFβ-induced microglial gene expression, thereby promoting the pro-tumorigenic microglia phenotype and glioma cell malignancy.
Sigma-Aldrich
TGF-beta 3-E. coli human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture