MilliporeSigma
  • Effect of plasma-derived extracellular vesicles on angiogenesis and the ensuing proliferative diabetic retinopathy through a miR-30b-dependent mechanism.
Effect of plasma-derived extracellular vesicles on angiogenesis and the ensuing proliferative diabetic retinopathy through a miR-30b-dependent mechanism.
Diabetology & metabolic syndrome (2022-12-10)
Ping Wang, Chengqian Li, Yujie Deng, Qing Yu, Xuxia Meng, Tao Jiang, Qing Wang, Yudong Fu
ABSTRACT
Proliferative diabetic retinopathy (PDR) is a major diabetic microvascular complication, characterized by pathological angiogenesis. This study sets out to investigate the potential molecular mechanism in the angiogenesis during PDR. The expression of microRNA-30b (miR-30b) was quantified in a streptozotocin (STZ)-induced mouse model of PDR. The binding affinity between SIRT1 and miR-30b was then identified and validated. After transduction with In-miR-30b or combined with sh-SIRT1, high-glucose (HG)-induced retinal microvascular endothelial cells (RMECs) were co-cultured with extracellular vesicles (EVs) derived from the plasma of PDR mice (plasma-EVs). The proliferation and angiogenesis of RMECs were then detected in vitro. miR-30b expression was upregulated in the retinal tissue of PDR mice. SIRT1 was a target gene of miR-30b and under the negative regulation by miR-30b in RMECs. In contrast, inhibition of miR-30b resulted in elevated SIRT1 expression, thus alleviating the angiogenesis of RMECs. miR-30b was enriched in the plasma-EVs and could be delivered to RMECs, in which miR-30b exerted pro-angiogenic effects. Furthermore, inhibition of miR-30b arrested the progression of PDR in mice by promoting the expression of SIRT1. Collectively, the present study pinpointed the involvement of miR-30b delivered by plasma-EVs in PDR angiogenesis, thus laying the basis for the development of novel therapeutic targets for the treatment of PDR.
Product Description
USP
Collagenase II, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
DAPI, for nucleic acid staining