MilliporeSigma
  • [3H]nisoxetine--a radioligand for noradrenaline reuptake sites: correlation with inhibition of [3H]noradrenaline uptake and effect of DSP-4 lesioning and antidepressant treatments.
[3H]nisoxetine--a radioligand for noradrenaline reuptake sites: correlation with inhibition of [3H]noradrenaline uptake and effect of DSP-4 lesioning and antidepressant treatments.
Neuropharmacology (1996-01-01)
S C Cheetham, J A Viggers, S A Butler, M R Prow, D J Heal
ABSTRACT
Nisoxetine is a potent and selective inhibitor of noradrenaline uptake into noradrenergic neurones. [3H]Nisoxetine binding to rat frontal cortical membranes was of high affinity. The binding data of both competition and saturation studies fitted a single site binding model. [3H]Nisoxetine binding was potently inhibited by the selective noradrenaline uptake inhibitors desipramine and protriptyline. In addition, a very good correlation was obtained between the ability of 25 monoamine reuptake inhibitors and related compounds both to inhibit [3H]nisoxetine binding and to inhibit [3H]noradrenaline uptake in rat frontal cortex. DSP-4 (10-100 mg/kg, i.p.) dose-dependently depleted cortical noradrenaline concentrations (51-100%), with no significant effects on 5-HT and dopamine. These depletions, which were used as a marker of loss of noradrenergic nerve terminals, were associated with a dose-dependent decrease in the number of [3H]nisoxetine binding sites (20-97%) with no change in binding affinity. Furthermore, a good correlation was obtained between cortical noradrenaline concentrations and the number of [3H]nisoxetine binding sites. These data support the view that [3H]nisoxetine binds to a single population of homogeneous sites associated with the noradrenaline transporter complex. Using this ligand, the effects of repeated administration of both antidepressant drugs with a range of pharmacological actions and of electroconvulsive shock on noradrenaline reuptake sites were examined. The number and affinity of [3H]nisoxetine binding sites were unaltered by all treatments. It is unlikely, therefore, that antidepressant therapy would produce adaptive changes in noradrenaline uptake sites.
Sigma-Aldrich
Nisoxetine hydrochloride, solid, ≥98% (HPLC)