MilliporeSigma
  • Cloning, expression, and characterization of a new beta-agarase from Vibrio sp. strain CN41.
Cloning, expression, and characterization of a new beta-agarase from Vibrio sp. strain CN41.
Applied and environmental microbiology (2011-08-09)
Li Liao, Xue-Wei Xu, Xia-Wei Jiang, Yi Cao, Na Yi, Ying-Yi Huo, Yue-Hong Wu, Xu-Fen Zhu, Xin-Qi Zhang, Min Wu
ABSTRACT
A new agarase, AgaA(CN41), cloned from Vibrio sp. strain CN41, consists of 990 amino acids, with only 49% amino acid sequence identity with known β-agarases. AgaA(CN41) belongs to the GH50 (glycoside hydrolase 50) family but yields neoagarotetraose as the end product. AgaA(CN41) was expressed and characterized.
Sigma-Aldrich
Agarase from Pseudomonas atlantica, lyophilized powder, ≥5,000 units/mg protein (Lowry)