MilliporeSigma
  • Complete genome sequence of the agarase-producing marine bacterium strain s89, representing a novel species of the genus Alteromonas.
Complete genome sequence of the agarase-producing marine bacterium strain s89, representing a novel species of the genus Alteromonas.
Journal of bacteriology (2011-09-15)
Chulhong Oh, Mahanama De Zoysa, Young-Kyung Kwon, Soo-Jin Heo, Abu Affan, Won-Kyo Jung, Heung-Sik Park, Jehee Lee, Seung-Kyu Son, Kon-Tak Yoon, Do-Hyung Kang
ABSTRACT
We report here the annotated genome sequence of the marine bacterium Alteromonas sp. S89 and the identification of six genes coding for agar-degrading enzymes. The sequenced Alteromonas sp. S89 genome is composed of a 3,864,871-bp circular chromosome that includes 3,236 complete open reading frames.
Sigma-Aldrich
Agarase from Pseudomonas atlantica, lyophilized powder, ≥5,000 units/mg protein (Lowry)