MilliporeSigma
  • Effects of manganese superoxide dismutase (MnSOD) expression on regulation of esophageal cancer cell growth and apoptosis in vitro and in nude mice.
Effects of manganese superoxide dismutase (MnSOD) expression on regulation of esophageal cancer cell growth and apoptosis in vitro and in nude mice.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine (2013-05-08)
Guogui Sun, Yadi Wang, Wanning Hu, Chengling Li
ABSTRACT
Manganese superoxide dismutase (MnSOD) catalyzes superoxide radical (O2 (-)) into hydrogen peroxide (H2O2), which is further catalyzed by the combined action of glutathione peroxidase (GPx) and catalase (CAT) into water and oxygen. MnSOD plays a role in cell protection from superoxide damage. This study aimed to investigate the effects of MnSOD on regulation of esophageal squamous cell carcinoma cell growth, apoptosis, and cell cycle distribution in vitro and tumor formation and growth in nude mouse xenografts. The data showed that differential levels of MnSOD expression had different effects on tumor cell proliferation, apoptosis, plating efficiency (PE), and cell cycle distribution in vitro and tumor formation and growth in nude mice. In particular, high levels of MnSOD expression promoted TE-1 cell growth and PE rate in vitro and in nude mice, whereas moderate MnSOD expression suppressed tumor cell growth and PE rate but induced more cell apoptosis. Thus, these data demonstrated the dual effects of MnSOD protein in esophageal squamous cell carcinoma and further study will confirm these current data.
Sigma-Aldrich
Superoxide Dismutase from bovine erythrocytes, BioUltra, lyophilized powder, ≥4,500 units/mg protein, ≥97% (SDS-PAGE)
Sigma-Aldrich
Superoxide Dismutase from bovine erythrocytes, lyophilized powder, ≥3,000 units/mg protein, Protein ≥95 % by biuret
Sigma-Aldrich
Superoxide Dismutase from bovine liver, ammonium sulfate suspension, 2,000-6,000 units/mg protein (biuret)
Sigma-Aldrich
Superoxide Dismutase from bovine erythrocytes, BioReagent, ≥3,000 units/mg protein, suitable for cell culture, lyophilized powder
Sigma-Aldrich
Superoxide Dismutase from bovine liver, lyophilized powder, ≥1500 units/mg protein
Sigma-Aldrich
Superoxide Dismutase bovine, recombinant, expressed in E. coli, lyophilized powder, ≥2500 units/mg protein, ≥90% (SDS-PAGE)
Sigma-Aldrich
Superoxide Dismutase from human erythrocytes, essentially salt-free, lyophilized powder, ≥2,500 units/mg protein