MilliporeSigma
  • Citrullination of proteins: a common post-translational modification pathway induced by different nanoparticles in vitro and in vivo.
Citrullination of proteins: a common post-translational modification pathway induced by different nanoparticles in vitro and in vivo.
Nanomedicine (London, England) (2012-05-26)
Bashir M Mohamed, Navin K Verma, Anthony M Davies, Aoife McGowan, Kieran Crosbie-Staunton, Adriele Prina-Mello, Dermot Kelleher, Catherine H Botting, Corey P Causey, Paul R Thompson, Ger Jm Pruijn, Elena R Kisin, Alexey V Tkach, Anna A Shvedova, Yuri Volkov
ABSTRACT
Rapidly expanding manufacture and use of nanomaterials emphasize the requirements for thorough assessment of health outcomes associated with novel applications. Post-translational protein modifications catalyzed by Ca(2+)-dependent peptidylargininedeiminases have been shown to trigger immune responses including autoantibody generation, a hallmark of immune complexes deposition in rheumatoid arthritis. Therefore, the aim of the study was to assess if nanoparticles are able to promote protein citrullination. Human A549 and THP-1 cells were exposed to silicon dioxide, carbon black or single-walled carbon nanotubes. C57BL/6 mice were exposed to respirable single-walled carbon nanotubes. Protein citrullination, peptidylargininedeiminases activity and target proteins were evaluated. The studied nanoparticles induced protein citrullination both in cultured human cells and mouse lung tissues. Citrullination occurred via the peptidylargininedeiminase-dependent mechanism. Cytokeratines 7, 8, 18 and plectins were identified as intracellular citrullination targets. Nanoparticle exposure facilitated post-translational citrullination of proteins.
Sigma-Aldrich
Peptidyl Arginine Deiminase from rabbit skeletal muscle, buffered aqueous glycerol solution, ≥200 units/mg protein (Bradford)
Sigma-Aldrich
Anti-Citrulline Antibody, Upstate®, from rabbit