MilliporeSigma
  • A tyrosinase gene missense mutation in temperature-sensitive type I oculocutaneous albinism. A human homologue to the Siamese cat and the Himalayan mouse.
A tyrosinase gene missense mutation in temperature-sensitive type I oculocutaneous albinism. A human homologue to the Siamese cat and the Himalayan mouse.
The Journal of clinical investigation (1991-03-01)
L B Giebel, R K Tripathi, R A King, R A Spritz
ABSTRACT
Type I oculocutaneous albinism (OCA) is an autosomal recessive disorder in which deficient synthesis of melanin pigment results from abnormal activity of melanocyte tyrosinase. A novel type I OCA phenotype in which hypopigmentation is related to local body temperature is associated with a missense substitution in tyrosinase, codon 422 CGG (Arg)----CAG (Gln). This substitution results in a tyrosinase polypeptide that is temperature-sensitive. This form of type I OCA thus is homologous to the temperature-related forms of albinism seen in the Siamese cat and the Himalayan mouse.
Sigma-Aldrich
Tyrosinase from mushroom, lyophilized powder, ≥1000 unit/mg solid