MilliporeSigma
  • Liquid chromatographic determination of aniline in table-top sweeteners based on pre-column derivatization with 1,2-naphthoquinone-4-sulfonate.
Liquid chromatographic determination of aniline in table-top sweeteners based on pre-column derivatization with 1,2-naphthoquinone-4-sulfonate.
Journal of chromatography. A (1999-11-26)
D Hernando, J Saurina, S Hernández-Cassou
ABSTRACT
A liquid chromatographic method for the determination of aniline in cyclamate sweeteners based on a pre-column derivatization with 1,2-naphthoquinone-4-sulfonate (NQS) is proposed. Aniline traces were extracted from the cyclamate samples using dichloromethane. After solvent evaporation, the dry residue was derivatized with NQS at pH 9.5 and 85 degrees C for 1 min. The aniline derivative, which was extracted from the reacting mixture, was redissolved in the eluent solution and injected into the chromatographic system. The separation of aniline derivative from other amine impurities was carried out in a C18 column using a 2% acetic acid-methanol (40:60, v/v) mobile phase. Results from the analysis of aniline in the sweetener samples with the proposed method were compared with those from the standard method. A good concordance between the two methods was observed.
Sigma-Aldrich
1,2-Naphthoquinone-4-sulfonic acid sodium salt, 97%