MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Gas chromatographic-mass spectrometric screening for organic acidemias using dried urine filter paper: determination of alpha-ketoacids.
Gas chromatographic-mass spectrometric screening for organic acidemias using dried urine filter paper: determination of alpha-ketoacids.
Journal of chromatography. B, Biomedical sciences and applications (2001-08-03)
X Fu, M Kimura, M Iga, S Yamaguchi
ABSTRACT
There are several organic acid disorders that require information on alpha-ketoacids, such as maple syrup urine disease or alpha-ketoadipic acidemia. The recovery, stability and diagnostic availability of alpha-ketoacids in dried urine filter paper analyzed by GC-MS with oxime-trimethylsilyl derivatization was studied for organic acidemia screening. The recovery of all nine types of alpha-ketoacids tested, but for phenylpyruvate, 2-ketoadipate, and p-OH-phenylpyruvate, from filter paper samples was acceptable. The stability of pyruvate, branched-chain alpha-ketoacids, alpha-ketoadipate and alpha-ketoglutarate was stable for at least 28 days, although some alpha-ketoacids such as succinylacetone were unstable. It indicated it was difficult to diagnose only tyrosinemia type 1 among nine specimens from organic acidemia patients tested. The method could be applied to global organic acidemia screening.
Sigma-Aldrich
2-Oxoadipic acid, ≥95.0% (HPLC)
Sigma-Aldrich
2-Oxovaleric acid, ≥98.0% (T)
Sigma-Aldrich
2-Hydroxy-2-methylbutyric acid, 98%