MilliporeSigma
  • Thrombostatin, a bradykinin metabolite, reduces platelet activation in a model of arterial wall injury.
Thrombostatin, a bradykinin metabolite, reduces platelet activation in a model of arterial wall injury.
Cardiovascular research (2002-03-30)
Alejandro R Prieto, Hongbao Ma, Ruiping Huang, Gauhar Khan, Kenneth A Schwartz, Elie E Hage-Korban, Alvin H Schmaier, John M Davis, Ahmed A K Hasan, George S Abela
ABSTRACT
Thrombin activates platelets and contributes to the occlusion of arteries following thrombolytic therapy or angioplasty. Thrombostatin (RPPGF), the angiotensin converting enzyme degradation product of bradykinin, inhibits alpha-thrombin induced platelet activation. We hypothesized that thrombostatin prevents platelet aggregation and adhesion after balloon angioplasty (BA). Platelet-rich plasma (PRP) was obtained from 22 Beagle dogs before sacrifice and 10% of the PRP was labeled with 111In. Carotid arteries were then removed from each dog and mounted in a dual perfusion chamber and intimal injury was performed with BA. 111In-PRP with or without thrombostatin or aspirin alone was perfused through the arteries for 60 min. During perfusion, platelet volume was measured using a Coulter counter and a laser-light scattering technique. Platelet adhesion to arteries was measured by radioactivity count. Arterial injury alone compared to non-injury increased platelet volume in the circuit by 1.4 times (x) (P<0.05) using a Coulter counter or 1.8x (P<0.05) using laser-light scattering and increased platelet adhesion by 2.3x (P<0.01). When compared to BA injury alone, the addition of thrombostatin reduced platelet volume by 1.8x (P<0.03) as measured by Coulter counter or 1.9x (P<0.01) by laser-light scattering and platelet adhesion by 4.2x (P<0.05). Compared to BA injury alone, aspirin reduced platelet volume by 1.2x (P<0.01) as assessed by Coulter counter or 1.5x (P<0.03) using laser-light scattering and platelet adhesion by 1.8x (P<0.02). Thrombostatin or aspirin independently decreases evidence of platelet activation in the canine carotid artery model of BA injury.
Sigma-Aldrich
Bradykinin Fragment 1-5, ≥97% (HPLC)