MilliporeSigma
  • Recent advances in synthetic transformations mediated by cerium(IV) ammonium nitrate.
Recent advances in synthetic transformations mediated by cerium(IV) ammonium nitrate.
Accounts of chemical research (2004-01-21)
Vijay Nair, Lakshmi Balagopal, Roshini Rajan, Jessy Mathew
ABSTRACT
Cerium(IV) ammonium nitrate (CAN) has recently emerged as a versatile reagent for oxidative electron transfer; the overwhelming number of reports serve as a testimony to the unparalleled utility of CAN in a variety of transformations of synthetic importance. Our recent work has uncovered novel carbon-carbon bond-forming reactions leading to the one-pot synthesis of dihydrofurans, tetrahydrofurans, and aminotetralins. In addition, we have developed a number of facile carbon-heteroatom bond-forming reactions by the CAN-mediated oxidative addition of soft anions to alkenes. A mechanistic rationale has been provided for the reactions explored. As might be expected of very powerful one-electron oxidants, the chemistry of cerium(IV) oxidation of organic molecules is dominated by radical and radical cation chemistry.
Sigma-Aldrich
Ammonium cerium(IV) nitrate, ≥99.99% trace metals basis
Sigma-Aldrich
Ammonium cerium(IV) nitrate, JIS special grade, ≥95.0%
Sigma-Aldrich
Ammonium cerium(IV) nitrate, puriss. p.a., ACS reagent, ≥98.5% (RT)
Sigma-Aldrich
Ammonium cerium(IV) nitrate, ≥98% (titration)
Sigma-Aldrich
Ammonium cerium(IV) nitrate, ACS reagent, ≥98.5%