MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Acaricidal activity of constituents identified in Foeniculum vulgare fruit oil against Dermatophagoides spp. (Acari: Pyroglyphidae).
Acaricidal activity of constituents identified in Foeniculum vulgare fruit oil against Dermatophagoides spp. (Acari: Pyroglyphidae).
Journal of agricultural and food chemistry (2004-05-13)
Hoi-Seon Lee
ABSTRACT
Acaricidal activities of components derived from Foeniculum vulgare fruit oil against Dermatophagoides farinae and Dermatophagoides pteronyssinus were examined using direct contact application and compared with that of the commercial repellent benzyl benzoate. The major biologically active constituent of Foeniculum fruit oil was characterized as (+)-fenchone by spectroscopic analyses. On the basis of LD(50) values, the compound most toxic to D. farinae was p-anisaldehyde (11.3 mg/m(2)) followed by (+)-fenchone (38.9 mg/m(2)), (-)-fenchone (41.8 mg/m(2)), benzyl benzoate (89.2 mg/m(2)), thymol (90.3 mg/m(2)), and estragol (413.3 mg/m(2)). Against D. pteronyssinus, p-anisaldehyde (10.1 mg/m(2)) was much more effective than benzyl benzoate (67.5 mg/m(2)), thymol (68.5 mg/m(2)), and estragol (389.9 mg/m(2)). These results indicate that the acaricidal activity of F. vulgare fruit oil likely results from (+)-fenchone and p-anisaldehyde. (+)-Fenchone was 20.3 times more abundant in the oil than p-anisaldehyde. (+)-Fenchone and p-anisaldehyde merit further study as potential house dust mite control agents or as lead compounds.
Sigma-Aldrich
L-Fenchone, ≥98%
Sigma-Aldrich
(1R)-(−)-Fenchone, ≥98%
Supelco
p-Anisaldehyde, analytical standard
Supelco
(+)-Fenchone, analytical standard
Sigma-Aldrich
(+)-Fenchone, purum, ≥98.0% (sum of enantiomers, GC)
Sigma-Aldrich
p-Anisaldehyde, 98%
Sigma-Aldrich
p-Anisaldehyde, ≥97.5%, FCC, FG