MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Nickel nitrate: a new junction permeability tracer for the study of the blood-testis barrier.
Nickel nitrate: a new junction permeability tracer for the study of the blood-testis barrier.
Microscopy research and technique (1992-01-01)
J C Cavicchia, F L Sacerdote, L A Gutierrez
ABSTRACT
With the purpose of evaluating a new intercellular tracer, nickel-K ferrocyanide, we compared results yielded by lanthanum with information provided by nickel. This was done in the seminiferous epithelium of Holtzman rats of several postnatal ages and in a wild local seasonal breeder Galea musteloides. Tissues were studied with transmission electron microscopy and freeze-fracture replications. Nickel tracing proved to delineate cell contours more intensely and less interruptedly than lanthanum. With regard to seasonal variations in adult galea, the limits of the barrier were similar to those described in other mammals: spermatogonia, preleptotene, and leptotene spermatocytes were surrounded by the tracer in the basal compartment. The zygotenepachytenes were contained in the lumenal compartment and tracers were stopped at the inter-Sertoli cell tight junctions. During the inactive spermatogenic phase in winter, the seminiferous epithelium contained Sertoli cells and occasional germ cells, never beyond the spermatocyte stage. The tracer filled intercellular spaces, indicating that the barrier was incompetent. Some resting germ cells showed nuclear hyperchromasia, karyolysis, organelle loss, cell shrinkage, and cell fusion leading to a multinucleated cells. The inter-Sertoli tight junctions were scanty and had randomly oriented and discontinuous junctional strands. Moreover, inter-Sertoli cell gap junctions proliferated. During the active spermatogenic phase in summer, junctions were numerous. Their junctional strands were parallel to each other, and continuous.
Sigma-Aldrich
Nickel(II) nitrate hexahydrate, puriss. p.a., ≥98.5% (KT)
Sigma-Aldrich
Nickel(II) nitrate hexahydrate, crystals
Sigma-Aldrich
Nickel(II) nitrate hexahydrate, purum p.a., crystallized, ≥97.0% (KT)
Sigma-Aldrich
Nickel(II) nitrate hexahydrate, 99.999% trace metals basis