MilliporeSigma
  • Adsorption and reactions of ClCH2CH2OH on clean and oxygen-precovered Cu(100): experimental and computational studies.
Adsorption and reactions of ClCH2CH2OH on clean and oxygen-precovered Cu(100): experimental and computational studies.
The journal of physical chemistry. B (2006-07-21)
Tao-Wei Fu, Yung-Hsuan Liao, Chia-Yuan Chen, Pei-Teng Chang, Ching-Yung Wang, Jong-Liang Lin
ABSTRACT
Temperature-programmed reaction/desorption, reflection-absorption infrared spectroscopy, and density functional theory calculations have been employed to investigate the adsorption and thermal reactions of ClCH2CH2OH on clean and oxygen-precovered Cu(100) surfaces. On Cu(100), ClCH2CH2OH is mainly adsorbed reversibly. The ClCH2CH2OH molecules at a submonolayer coverage can change their orientation with increasing temperature. However, on oxygen-precovered Cu(100), all of the adsorbed ClCH2CH2OH molecules below 0.5 langmuir exposures completely dissociate to generate ethylene and acetaldehyde via the intermediate of ClCH2CH2O-. The computational studies predict that the ClCH2CH2O- is most likely to be adsorbed at the 4-fold hollow sites of Cu(100), with its C-O bond only slightly titled away from the surface normal and with a gauche conformation with respect to the C-C bond. The hollow-site ClCH2CH2O- has an adsorption energy that is 4.4 and 19.2 kcal x mol(-1) lower than that of the ClCH2CH2O- bonded at the bridging and atop sites, respectively. No significant effect of precovered oxygen on the ClCH2CH2O- bonding geometry and infrared band frequencies has been observed, as compared with the case without oxygen.
Sigma-Aldrich
2-Chloroethanol, SAJ special grade, ≥99.0%
Sigma-Aldrich
2-Chloroethanol, 99%