MilliporeSigma
  • Development of indole chemistry to label tryptophan residues in protein for determination of tryptophan surface accessibility.
Development of indole chemistry to label tryptophan residues in protein for determination of tryptophan surface accessibility.
Protein science : a publication of the Protein Society (2007-05-26)
Carol L Ladner, Raymond J Turner, Robert A Edwards
ABSTRACT
Solvent accessibility can be used to evaluate protein structural models, identify binding sites, and characterize protein conformational changes. The differential modification of amino acids at specific sites enables the accessible surface residues to be identified by mass spectrometry. Tryptophan residues within proteins can be differentially labeled with halocompounds by a photochemical reaction. In this study, tryptophan residues of carbonic anhydrase are reacted with chloroform, 2,2,2-trichloroethanol (TCE), 2,2,2-trichloroacetate (TCA), or 3-bromo-1-propanol (BP) under UV irradiation at 280 nm. The light-driven reactions with chloroform, TCE, TCA, and BP attach a formyl, hydroxyethanone, carboxylic acid, and propanol group, respectively, onto the indole ring of tryptophan. Trypsin and chymotrypsin digests of the modified carbonic anhydrase are used to map accessible tryptophan residues using matrix-assisted laser desorption/ionization time-of-flight mass spectrometry (MALDI-TOF MS). Tryptophan reactivity is determined by identifying peptides with tryptophan residues modified with the appropriate label. The reactivity is calculated from the frequency that the modification is identified and a semiquantitative measure of the amount of products formed. Both of these measures of tryptophan reactivity correlate significantly with the accessible surface area of tryptophan residues in carbonic anhydrase determined from the X-ray crystal structure. Therefore the photochemical reaction of halocompounds with tryptophan residues in carbonic anhydrase indicates the degree of solvent accessibility of these residues.
Sigma-Aldrich
2-Chloroethanol, SAJ special grade, ≥99.0%
Sigma-Aldrich
2-Chloroethanol, 99%
Sigma-Aldrich
2,2,2-Trichloroethanol, ReagentPlus®, ≥99%