MilliporeSigma
  • An efficient solvent-free synthesis of benzoxanthenes catalyzed by BSA or O-TSA.
An efficient solvent-free synthesis of benzoxanthenes catalyzed by BSA or O-TSA.
Preparative biochemistry & biotechnology (2008-06-24)
G Luo, D Liu, C Liu
ABSTRACT
A series of 14-alkyl and aryl-14H-dibenzo[a.j]xanthenes have been synthesized with high yields through a one-pot condensation of beta-naphthol with alkyl or aryl aldehydes in the presence of BSA or o-TSA in a solvent-free medium.
Sigma-Aldrich
p-Toluenesulfonamide, reagent grade, 97%
Sigma-Aldrich
p-Toluenesulfonamide, ReagentPlus®, ≥99%