MilliporeSigma
  • Matrix metaloproteinases activity during the evolution of hypoxic-ischemic brain damage in the immature rat. The effect of 1-methylnicotinamide (MNA).
Matrix metaloproteinases activity during the evolution of hypoxic-ischemic brain damage in the immature rat. The effect of 1-methylnicotinamide (MNA).
Journal of physiology and pharmacology : an official journal of the Polish Physiological Society (2008-10-28)
P Dragun, D Makarewicz, L Wójcik, M Ziemka-Nałecz, M Słomka, T Zalewska
ABSTRACT
Matrix metalloproteinases (MMPs) are a family of proteolytic enzymes that degrade the extracellular matrix and carry out key functions during brain development. Apart from a physiological role, excessive activation of MMPs in brain tissue has been postulated to represent a pathway for cell death arising from ischemia. To evaluate the possible involvement of MMPs in the perinatal brain asphyxia, we exposed 7-day-old rats to hypoxia-ischemia (HI). Unilateral HI was administered by ligation of the common carotid artery followed by hypoxia (7.4% O2/92.6% N2) for 65 minutes. This insult is known to produce brain damage confined to the cerebral hemisphere ipsilateral to the arterial occlusion in > 90% of animals. HI resulted in a significant elevation of MMP-2 and MMP-9 activity in the ipsilateral forebrain. The maximum activation was found at 48 hours and 7-14 days after the insult. These results suggest that early and late induction of MMPs may play a role in neuronal death as well as in repair processes. The treatment of animals subjected to HI with 1-methylnicotinamide (MNA), the anti-inflammatory agent, led to the inhibition of MMP-9 in an acute phase of ischemic damage and to the activation of MMP-2 in the later stages after injury. The timing of MMPs modulation by MNA may indicate its possible therapeutic implications.
Product Description
Supelco
1-Methylnicotinamide chloride, analytical standard