MilliporeSigma
New antitumour fungal metabolites from Alternaria porri.
Natural product research (2009-08-08)
Preecha Phuwapraisirisan, Jakaphan Rangsan, Pongpan Siripong, Santi Tip-Pyang
ABSTRACT
Chemical investigation of the onion pathogenic fungus Alternaria porri resulted in the isolation of two new phthalides named zinnimide (2) and deprenylzinnimide (8), along with a new bianthraquinone, alterporriol F (10). The structures of the new metabolites were characterised by spectroscopic analysis and chemical degradation. Of the new compounds isolated, alterporriol F was highly cytotoxic towards HeLa and KB cells, with IC(50) values of 6.5 and 7.0 microg mL(-1).
Sigma-Aldrich
Alternariol from Alternaria sp., ~96%
Supelco
Alternariol, analytical standard