MilliporeSigma
  • Validation of a liquid chromatographic/tandem mass spectrometric method for the determination of scopolamine butylbromide in human plasma: application of the method to a bioequivalence study.
Validation of a liquid chromatographic/tandem mass spectrometric method for the determination of scopolamine butylbromide in human plasma: application of the method to a bioequivalence study.
Journal of AOAC International (2009-11-18)
Josélia Larger Manfio, Mauricio Bedim Dos Santos, Wagner Alex Jann Favreto, Fabiane Ines Hoffmann, Adriana Cristina Mertin
ABSTRACT
A sensitive and specific LC/MS/MS method was developed and validated for the determination of scopolamine butylbromide in human plasma. Scopolamine butylbromide and propanolol (internal standard) were extracted from the plasma by liquid-liquid extraction with dichloromethane as the extraction solvent and separated on a C18 analytical column (50 x 4.6 mm id) maintained at 40 degrees C. The analytes were eluted at a constant flow rate of 0.45 mL/min; the mobile phase consisted of acetonitrile and a buffer of 5 mM ammonium acetate and 0.1% formic acid (60 + 40, v/v). The mass spectrometer, equipped with an electrospray source in the positive ionization mode, was set up in the multiple-reaction monitoring mode to monitor the transitions m/z 360.6 > 102.5 (scopolamine butylbromide) and m/z 259.7 > 115.6 (propanolol). The chromatographic separation was obtained within 2.0 min, and the responses were linear over the concentration range of 0.10-40.00 ng/mL. The mean extraction recoveries of scopolamine butylbromide and propanolol from plasma were 69.00 and 80.76%, respectively. Method validation parameters, such as specificity, linearity, precision, accuracy, and stability, were within the acceptable range. Moreover, when the proposed method was successfully applied to a pharmacokinetic study of healthy human volunteers, the results showed that the two scopolamine butylbromide formulations tested are not bioequivalent in rate and extent of absorption.
Product Description
Supelco
1-Bromobutane, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
1-Bromobutane, ReagentPlus®, 99%
Pricing and availability is not currently available.