MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Comparative in vitro safety analysis of dyes for chromovitrectomy: indocyanine green, brilliant blue green, bromophenol blue, and infracyanine green.
Comparative in vitro safety analysis of dyes for chromovitrectomy: indocyanine green, brilliant blue green, bromophenol blue, and infracyanine green.
Retina (Philadelphia, Pa.) (2011-03-12)
Sankarathi Balaiya, Vikram S Brar, Ravi K Murthy, Kakarla V Chalam
ABSTRACT
Vital dyes such as infracyanine green (IfCG), brilliant blue green (BBG), and bromophenol blue (BPB) have been used as an alternative to indocyanine green (ICG) during chromovitrectomy. We compared the in vitro toxicity of IfCG, BBG, and BPB with ICG on the retinal pigment epithelial cells and retinal ganglion cells at various concentrations to optimize the safe dose and duration of exposure. Cultured retinal ganglion cells (RGC-5) and human retinal pigment epithelial cells (ARPE-19) were exposed to 2 concentrations (0.25 and 0.5 mg/mL) of ICG, IfCG, BBG, and BPB at various time intervals (1, 5, 15, and 30 minutes). Cell viability was quantified with neutral red assay, and mode of cell death was evaluated with flow cytometry-based Annexin V and propidium iodide staining. Exposure to ICG resulted in 48%-74% reduction in neutral red uptake in both RGC-5 and ARPE-19 cells, after an exposure time of ≥5 minutes compared with control (P < 0.001). Infracyanine green, BBG, and BPB were significantly less toxic on the 2 cell lines at exposure times <15 minutes. (Reduction in cell viability ranged from 6.9% ± 3.3% to 29.3% ± 7.4% when compared with control, P > 0.5.) However, among the newer dyes, BBG caused necrosis in retinal pigment epithelial cells and retinal ganglion cells as the exposure time period increased beyond 5 minutes. Newer vital dyes, IfCG, BBG, and BPB, are significantly less toxic on retinal ganglion cells and retinal pigment epithelial cells' cell lines when compared with ICG. Infracyanine green was least toxic among the three newer dyes studied.
Sigma-Aldrich
Bromophenol Blue, JIS special grade
Sigma-Aldrich
Bromophenol Blue, titration: suitable
Sigma-Aldrich
Bromophenol Blue, ACS reagent