MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Mild decarboxylative activation of malonic acid derivatives by 1,1'-carbonyldiimidazole.
Mild decarboxylative activation of malonic acid derivatives by 1,1'-carbonyldiimidazole.
Organic letters (2011-04-15)
Danny Lafrance, Paul Bowles, Kyle Leeman, Robert Rafka
ABSTRACT
Malonic acid derivatives undergo unusually mild decarboxylation when treated with N,N'-carbonyldiimidazole (CDI) at room temperature to generate the carbonyl imidazole moiety in high yield, which can be reacted further with a variety of nucleophiles in an efficient one-pot process.
Sigma-Aldrich
CDI, ≥97.0% (T)
Sigma-Aldrich
CDI, reagent grade