MilliporeSigma
  • Effect of different surface treatments on the repair strength of a nanofilled resin-based composite.
Effect of different surface treatments on the repair strength of a nanofilled resin-based composite.
Dental materials journal (2011-07-23)
Naho Hamano, Yu-Chih Chiang, Indra Nyamaa, Hiroaki Yamaguchi, Satoshi Ino, Reinhard Hickel, Karl-Heinz Kunzelmann
ABSTRACT
The purpose of our study was to re-evaluate the effects of surface treatment on the bond strength of repaired methacrylate-based dental composite materials including nanofillers (Ceram X, Dentsply). The microtensile bond strengths were measured before or after thermo-mechanical fatigue simulation. The treatments were as follows: (1)No treatment (negative control), (2)Bonding agent, (3) Acetone, (4)Acetone+Bonding agent, (5)Silane, (6)Silane+Bonding agent. In the digitized SEM image of the polished aged specimens without any surface treatment, the percentage of the visible fillers relative to the whole image was calculated. Before and after fatigue, no significant differences could be observed among the different pretreatment groups. Fatigue increased the microtensile strength values of Acetone and Silane groups. The digitized SEM image reveals that 11.5% of the exposed surface could be identified as uncovered filler surface. None of surface treatments (acetone, silane and bonding agent) contributed to increase the tensile strength of repaired composite samples including nanofillers.
Sigma-Aldrich
3-(Trimethoxysilyl)propyl methacrylate, 98%
Sigma-Aldrich
3-(Trimethoxysilyl)propyl methacrylate, ≥97%