MilliporeSigma
  • Regulation of NAD(P)H dehydrogenase-dependent cyclic electron transport around PSI by NaHSO₃ at low concentrations in tobacco chloroplasts.
Regulation of NAD(P)H dehydrogenase-dependent cyclic electron transport around PSI by NaHSO₃ at low concentrations in tobacco chloroplasts.
Plant & cell physiology (2011-08-11)
Yanxia Wu, Fangfang Zheng, Weimin Ma, Zhiguo Han, Qun Gu, Yunkang Shen, Hualing Mi
ABSTRACT
Although bisulfite at low concentrations (L-NaHSO₃) has been found to increase the cyclic electron transport around PSI (CET), its regulative mechanism remains unknown. In this work, the role of L-NaHSO₃ (0.1-500 μM) in NAD(P)H dehydrogenase-dependent CET (the NDH pathway) was investigated. After treatment of tobacco leaves with L-NaHSO₃, the NDH pathway, as reflected by a transient post-illumination increase in Chl fluorescence, the dark reduction of P700+ after far-red light and the amount of NDH, was increased after the light-dark-light transition, but was slightly lowered under continuous light. Meanwhile, the linear electron transport (LET) was accelerated by L-NaHSO₃ under both the light regimes. Experiments in thylakoids further demonstrated that both LET, monitored by light-dependent oxygen uptake, and CET, as determined from the NADPH-dependent oxygen uptake and dark reduction of P700+, were enhanced by L-NaHSO₃ and the enhancements were abolished by superoxide dismutase. Furthermore, L-NaHSO₃-induced CET was partially impaired in thylakoids of the ΔndhCKJ mutant, while L-NaHSO₃-induced LET was not affected. Based on these results, we propose that the photooxidation of L-NaHSO₃ initiated by superoxide anions in PSI regulates NDH pathway to maintain efficient photosynthesis.
Sigma-Aldrich
Sodium bisulfite, JIS special grade, 64.0-67.4% SO2 basis
Sigma-Aldrich
Sodium bisulfite, anhydrous, free-flowing, Redi-Dri, ACS reagent
Sigma-Aldrich
Sodium bisulfite, ACS reagent
Sigma-Aldrich
Sodium bisulfite solution, purum, ~40%