MilliporeSigma
  • Photoactivation studies of zinc porphyrin-myoglobin system and its application for light-chemical energy conversion.
Photoactivation studies of zinc porphyrin-myoglobin system and its application for light-chemical energy conversion.
International journal of biological sciences (2011-11-02)
Chin-Hao Chang, Yi-Ting Hu, Chen-Fu Lo, Liyang Luo, Hung-Ming Lin, Cheng-Hsiang Chang, Ching-Yao Lin, Eric Wei-Guang Diau, Tung-Kung Wu
ABSTRACT
An artificial zinc porphyrin-myoglobin-based photo-chemical energy conversion system, consisting of ZnPP-Mb or ZnPE(1)-Mb as a photosensitizer, NADP(+) as an electron acceptor, and triethanolamine as an electron donor, has been constructed to mimic photosystem I. The photoirradiated product is able to reduce a single-electron acceptor protein cytochrome c, but cannot catalyze the two-electron reduction of acetaldehyde by alcohol dehydrogenase, thus demonstrating a single electron transfer mechanism. Furthermore, the artificial system can bifunctionally promote oxidoredox reactions, depending on the presence or absence of a sacrificial electron donor, thus suggesting its potential application in electrochemical regeneration steps involved in chemical transformation and/or energy conversion.
Sigma-Aldrich
Triethanolamine hydrochloride, BioXtra, ≥99.5% (titration)
Sigma-Aldrich
Triethanolamine hydrochloride, ≥99.5% (titration)
Sigma-Aldrich
Triethanolamine, ≥99.0% (GC)
Sigma-Aldrich
Triethanolamine, puriss. p.a., ≥99% (GC)
Sigma-Aldrich
Triethanolamine, puriss., meets analytical specification of NF, ≥99% (GC)
Sigma-Aldrich
Triethanolamine, JIS special grade, ≥98.0%
Sigma-Aldrich
Triethanolamine, reagent grade, 98%
Sigma-Aldrich
Triethanolamine, BioUltra, ≥99.5% (GC)