MilliporeSigma
  • N-Acetyltransferase 2 genotype, exfoliated urothelial cells and benzidine exposure.
N-Acetyltransferase 2 genotype, exfoliated urothelial cells and benzidine exposure.
Frontiers in bioscience (Elite edition) (2011-12-29)
Qing-wen Ma, Guo-fang Lin, Ji-gang Chen, Wei-Chao Guo, Yi-qiu Qin, Klaus Golka, Jian-hua Shen
ABSTRACT
Most studies report an association of the slow N-acetyltransferase 2 (NAT2) status with elevated bladder cancer risk. In this study, NAT2 genotypes and the decades-long records of Papanicolaou's grading of exfoliated urothelial cells in a former benzidine-exposed cohort of the Shanghai dyestuff industry (29 bladder cancer patients; 307 non-cancer cohort members, some of them presenting different grades of pre-malignant alterations of exfoliated urothelial cells) were investigated. The cohort members had been enrolled in regular medical surveillance since mid-1980s. No overall increase of slow NAT2 genotypes in the former benzidine-exposed bladder cancer patients was found, compared with non-diseased members of the same cohort. A lower presentation of the homozygous wild genotype NAT2 4/4 was observed in bladder cancer patients, compared with non-diseased members with averaged Papanicolaou's grading (APG)3 II (OR=0.31, 95 percent CI 0.10-0.96, p=0.034) or with APG less than II (OR=0.36,95 percent CI 0.12-1.10, p=0.063). Nevertheless, neither a protective influence of rapid NAT2 genotypes on bladder cancer risk nor on pre-malignant cytological alterations could be confirmed by the present data.
Product Description
Supelco
Benzidine, analytical standard
Sigma-Aldrich
Benzidine, ≥98%
Sigma-Aldrich
Benzidine dihydrochloride, ≥99% (titration)