- Synthesis of 4-methylcoumarin derivatives containing 4,5-dihydropyrazole moiety to scavenge radicals and to protect DNA.
Synthesis of 4-methylcoumarin derivatives containing 4,5-dihydropyrazole moiety to scavenge radicals and to protect DNA.
European journal of medicinal chemistry (2012-04-24)
Chuan Xiao, Xu-Yang Luo, De-Jun Li, Hang Lu, Zai-Qun Liu, Zhi-Guang Song, Ying-Hua Jin
PMID22521371
ABSTRACT
A series of 4-methylcoumarin derivatives containing 4,5-dihydropyrazole moiety were synthesized and their antioxidant activities were evaluated in AAPH (2,2'-azobis(2-amidinopropane hydrochloride))-induced oxidation of DNA, and in trapping DPPH (2,2'-diphenyl-1-picrylhydrazyl) and ABTS(+•) (2,2'-azinobis(3-ethylbenzothiazoline-6-sulfonate) cationic radical), respectively. Among coumarin derivatives, 3a-d and 4a-c exhibited the termination of radical propagation-chains in AAPH-induced oxidation of DNA. The ortho dihydroxyphenyl substitution at 5 position and 1-unsubstitution of the 4,5-dihydroxylpyrazole was found enhancing the antioxidant activities of these coumarin derivatives.