MilliporeSigma
  • Pseudo-enzymatic hydrolysis of 4-nitrophenyl acetate by human serum albumin: pH-dependence of rates of individual steps.
Pseudo-enzymatic hydrolysis of 4-nitrophenyl acetate by human serum albumin: pH-dependence of rates of individual steps.
Biochemical and biophysical research communications (2012-07-10)
Paolo Ascenzi, Magda Gioia, Gabriella Fanali, Massimo Coletta, Mauro Fasano
ABSTRACT
Human serum albumin (HSA) displays esterase activity reflecting multiple irreversible chemical modifications rather than turnover. Here, kinetics of the pseudo-enzymatic hydrolysis of 4-nitrophenyl acetate (NphOAc) are reported. Under conditions where [HSA]≥ 5×[NphOAc] and [NphOAc]≥ 5×[HSA], the HSA-catalyzed hydrolysis of NphOAc is a first-order process for more than 95% of its course. From the dependence of the apparent rate constants k(app) and k(obs) on [HSA] and [NphOAc], respectively, values of K(s), k(+2), and k(+2)/K(s) were determined. Values of K(s), k(+2), and k(+2)/K(s) obtained at [HSA]≥ 5×[NphOAc] and [NphOAc]≥ 5×[HSA] are in good agreement, the deacylation step being rate limiting in catalysis. The pH-dependence of k(+2)/K(s), k(+2), and K(s) reflects the acidic pK(a) shift of the Tyr411 catalytic residue from 9.0 ± 0.1 in the substrate-free HSA to 8.1 ± 0.1 in the HSA:NphOAc complex. Accordingly, diazepam inhibits competitively the HSA-catalyzed hydrolysis of NphOAc by binding to Tyr411.
Sigma-Aldrich
4-Nitrophenyl acetate, esterase substrate