MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Correlation between surface roughness and microhardness of experimental composites with varying filler concentration.
Correlation between surface roughness and microhardness of experimental composites with varying filler concentration.
The journal of contemporary dental practice (2012-08-25)
Eliseu Aldrighi Munchow, Marcos Brito Correa, Fabricio Aulo Ogliari, Evandro Piva, Cesar Henrique Zanchi
ABSTRACT
The purpose of this study was to investigate the influence of the surface roughness on the surface microhardness of experimental composites with varying filler concentration. Experimental resin composites were formulated by mixing Bis-GMA and TEGDMA in a 50/50% weight ratio and CQ/EDAB were added to make the material photosensitive. Silanized glass particles were incorporated in the resin blend in two concentrations: C50 with 50% and C75 with 75% in weight ratio. The surface roughness and the surface microhardness measurements were determined after every three finishing procedures with #280-, #600- and #1200-grit wet sandpapers, respectively. The data were analyzed statistically by Two Way ANOVA and Tukey's test, and comparisons were conducted using the Spearman's correlation test (p > 0.05). The surface roughness and surface microhardness were negatively associated (r = - 0.68) and the finishing procedures of both composites resulted in harder and smoother surfaces than the initial ones. Additionally, in a smooth circumstance, the higher content of fillers has not resulted in a composite with better microhardness and smoothness. Finishing procedures decreased the surface roughness and consequently improved the surface microhardness of the composites evaluated. Finishing and polishing procedures are effectives in reducing the surface roughness amplitude of composite materials and in improving their surface microhardness. Thus a microhardness test and any hardness evaluation must be conducted only after a properly finished and polished surface is achieved.
Sigma-Aldrich
Triethylene glycol dimethacrylate, contains 200 ppm monomethyl ether hydroquinone as inhibitor, 99%
Sigma-Aldrich
Triethylene glycol dimethacrylate, contains 80-120 ppm MEHQ as inhibitor, 95%