MilliporeSigma
  • Ethylenediamine diacetate (EDDA) mediated synthesis of aurones under ultrasound: their evaluation as inhibitors of SIRT1.
Ethylenediamine diacetate (EDDA) mediated synthesis of aurones under ultrasound: their evaluation as inhibitors of SIRT1.
Bioorganic & medicinal chemistry letters (2012-08-30)
Khanapur Manjulatha, S Srinivas, Naveen Mulakayala, D Rambabu, M Prabhakar, Kalle M Arunasree, Mallika Alvala, M V Basaveswara Rao, Manojit Pal
ABSTRACT
An improved synthesis of functionalized aurones has been accomplished via the reaction of benzofuran-3(2H)-one with a range of benzaldehydes in the presence of a mild base EDDA under ultrasound. A number of aurones were synthesized (within 5-30min) and the molecular structure of a representative compound determined by single crystal X-ray diffraction study confirmed Z-geometry of the C-C double bond present within the molecule. Some of the compounds synthesized have shown SIRT1 inhibiting as well as anti proliferative properties against two cancer cell lines in vitro. Compound 3a [(Z)-2-(5-bromo-2-hydroxybenzylidene) benzofuran-3(2H)-one] was identified as a potent inhibitor of SIRT1 (IC(50)=1μM) which showed a dose dependent increase in the acetylation of p53 resulting in induction of apoptosis.
Sigma-Aldrich
Ethylenediamine dihydrochloride, 98%
Sigma-Aldrich
Ethylenediamine diacetate, 98%
Sigma-Aldrich
Ethylenediamine, SAJ special grade, ≥99.0%
Sigma-Aldrich
Ethylenediamine, meets USP testing specifications
Sigma-Aldrich
Ethylenediamine, puriss. p.a., absolute, ≥99.5% (GC)
Sigma-Aldrich
Ethylenediamine, ReagentPlus®, ≥99%
Sigma-Aldrich
Ethylenediamine, purified by redistillation, ≥99.5%
Sigma-Aldrich
Ethylenediamine, BioXtra
Sigma-Aldrich
Ethylenediamine solution, technical, 75-80%
Supelco
Ethylenediamine, analytical standard