MilliporeSigma
Highly ion-conducting poly(ionic liquid) layers.
Chemical communications (Cambridge, England) (2012-09-11)
Thomas J Wood, Wayne C E Schofield, Peter Lund, Mikkel J Larsen, Jas Pal S Badyal
ABSTRACT
Highly ion-conducting poly(ionic liquid) thin films have been prepared in the absence of solvents utilizing a simple 2-step approach comprising pulsed plasmachemical deposition of 1-allylimidazole followed by vapour-phase quaternization with 1-bromobutane.
Sigma-Aldrich
1-Bromobutane, ReagentPlus®, 99%
Pricing and availability is not currently available.
Supelco
1-Bromobutane, analytical standard
Pricing and availability is not currently available.