MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Comparative analysis of the main bioactive components of San-ao decoction and its series of formulations.
Comparative analysis of the main bioactive components of San-ao decoction and its series of formulations.
Molecules (Basel, Switzerland) (2012-11-03)
Xiaoyun Shu, Yuping Tang, Chenxue Jiang, Erxing Shang, Xinshen Fan, Anwei Ding
ABSTRACT
A high performance liquid chromatographic (HPLC) method with diode array detection (DAD) was established for simultaneous determination of seven main bioactive components in San-ao decoction and its series of formulae (San-ao decoction, Wu-ao decoction, Qi-ao decoction and Jia-wei San-ao decoction). Seven compounds were analyzed simultaneously with a XTerra C(18) column (4.6 mm × 250 mm, 5 µm) using a linear gradient elution of a mobile phase containing acetonitrile (A) and a buffer solution (0.02 mol/L potassium dihydrogen phosphate and adjusted to pH 3 using phosphoric acid) (B); the flow rate was 1.0 mL/min. The sample was detected with DAD at 210, 254 and 360 nm and the column was maintained at 30 °C. All the compounds showed good linearity (r2 > 0.9984) in the tested concentration range. The precisions were evaluated by intra-day and inter-day tests, and relative standard deviation (R.S.D.) values within the range of 0.83%–2.53% and 0.64%–2.77% were reported, respectively. The recoveries of the quantified compounds were observed to cover a range from 95.34% and 104.82% with R.S.D. values less than 2.72%. The validated method was successfully applied for the simultaneous determination of seven main bioactive components including ephedrine (1), amygdalin (2), liquiritin (3), benzoic acid (4), isoliquiritin (5), formononetin (6) and glycyrrhizic acid (7) in San-ao decoction and its series of formulae. The results also showed a wide variation in the content of the identified active compounds in these samples, which could also be helpful to illustrate the drug interactions after some herbs combined in different formulations.
Sigma-Aldrich
Amygdalin, BioXtra, ≥97.0% (HPLC)
Sigma-Aldrich
Amygdalin, ≥99% (HPLC), from apricot kernels
Sigma-Aldrich
Formononetin, ≥99.0% (TLC)
Supelco
Mettler-Toledo Calibration substance ME 18555, Benzoic acid, analytical standard, (for the calibration of the melting point system), traceable to primary standards (LGC)
Supelco
Benzoic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Benzoic acid, SAJ first grade, ≥99.5%
Sigma-Aldrich
(1R,2S)-(−)-Ephedrine, 98%
Supelco
Benzoic acid, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Benzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
Benzoic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Benzoic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9% (alkalimetric)
Sigma-Aldrich
Benzoic acid, purified by sublimation, ≥99%
Supelco
Benzoic acid, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Benzoic acid, meets analytical specification of Ph. Eur., BP, USP, FCC, E210, 99.5-100.5% (alkalimetric)
Supelco
Melting point standard 121-123°C, analytical standard
Sigma-Aldrich
Potassium benzoate, ReagentPlus®, 99%
Sigma-Aldrich
Benzoic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Benzoic acid, SAJ special grade, ≥99.5%
Sigma-Aldrich
Benzoic acid, natural, ≥99.5%, FCC, FG