MilliporeSigma
  • Luteolin induces carcinoma cell apoptosis through binding Hsp90 to suppress constitutive activation of STAT3.
Luteolin induces carcinoma cell apoptosis through binding Hsp90 to suppress constitutive activation of STAT3.
PloS one (2012-11-13)
Jin Fu, Dan Chen, Bo Zhao, Zhihui Zhao, Jiahong Zhou, Yimiao Xu, Yinqiang Xin, Chang Liu, Lan Luo, Zhimin Yin
ABSTRACT
Abnormal activity of STAT3 is associated with a number of human malignancies. Hsp90 plays a central role in stabilizing newly synthesized proteins and participates in maintaining the functional competency of a number of signaling transducers involved in cell growth, survival and oncogenesis, such as STAT3. Hsp90 interacts with STAT3 and stabilizes Tyr-phosphorylated STAT3. It has been reported that luteolin possesses anticancer activity through degradation of Tyr(705)-phosphorylated STAT3. We found that overexpression of Hsp90 inhibited luteolin-induced degradation of Tyr(705)-phosphorylated STAT3 and luteolin also reduced the levels of some other Hsp90 interacting proteins. Results from co-immunoprecipitation and immunoblot analysis demonstrated that luteolin prevented the association between Hsp90 and STAT3 and induced both Tyr(705)- and Ser(727)-phosphorylated STAT3 degradation through proteasome-dependent pathway. The molecular modeling analysis with CHARMm-Discovery Studio 2.1(DS 2.1) indicated that luteolin could bind to the ATP-binding pocket of Hsp90. SPR technology-based binding assay confirmed the association between luteolin and Hsp90. ATP-sepharose binding assay displayed that luteolin inhibited Hsp90-ATP binding. Luteolin promoted the degradation of Tyr(705)- and Ser(727)-phosphorylated STAT3 through interacting with Hsp90 and induced apoptosis of cancer cells. This study indicated that luteolin may act as a potent HSP90 inhibitor in antitumor strategies.
Sigma-Aldrich
Caspase Inhibitor VI, Z-VAD-FMK, CAS 161401-82-7, is an irreversible pan caspase inhibitor. Does not require pretreatment with esterase for in vitro studies.
Sigma-Aldrich
Luteolin, ≥98% (TLC), powder
Supelco
Luteolin, analytical standard
Luteolin, primary reference standard