MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Simulation of the plant uptake of organophosphates and other emerging pollutants for greenhouse experiments and field conditions.
Simulation of the plant uptake of organophosphates and other emerging pollutants for greenhouse experiments and field conditions.
Environmental science and pollution research international (2012-12-06)
Stefan Trapp, Trine Eggen
ABSTRACT
The uptake of the organophosphates tris(2-chloroethyl) phosphate (TCEP), tris(1-chloro-2-propyl) phosphate (TCPP), tributyl phosphate (TBP), the insect repellant N,N-diethyl toluamide (DEET), and the plasticizer n-butyl benzenesulfonamide (NBBS) into plants was studied in greenhouse experiments and simulated with a dynamic physiological plant uptake model. The calibrated model was coupled to a tipping buckets soil transport model and a field scenario with sewage sludge application was simulated. High uptake of the polar, low-volatile compounds TCEP, TCPP, and DEET into plants was found, with highest concentrations in straw (leaves and stem). Uptake into carrot roots was high for TCPP and TBP. NBBS showed no high uptake but was rapidly degraded. Uptake into barley seeds was small. The pattern and levels of uptake could be reproduced by the model simulations, which indicates mainly passive uptake and transport (i.e., by the transpiration stream, with the water) into and within the plants. Also the field simulations predicted a high uptake from soil into plants of TCEP, TCPP, and DEET, while TBP is more likely taken up from air. The BCF values measured and calculated in the greenhouse study are in most cases comparable to the calculated values of the field scenario, which demonstrates that greenhouse studies can be suitable for predicting the behavior of chemicals in the field. Organophosphates have a high potential for bioaccumulation in crops and reach agricultural fields both via sewage sludge and by atmospheric deposition.
Sigma-Aldrich
N-Butylbenzenesulfonamide, 99%
Sigma-Aldrich
N,N-Diethyl-3-methylbenzamide, 97%
Sigma-Aldrich
4,4′,4′′,4′′′-(Porphine-5,10,15,20-tetrayl)tetrakis(benzoic acid), Dye content 75 %
Supelco
DEET, PESTANAL®, analytical standard
Sigma-Aldrich
Tributyl phosphate, puriss., ≥99.0% (GC)
Supelco
Tributyl phosphate, for extraction analysis, ≥99.0% (GC)
Sigma-Aldrich
Tributyl phosphate, ≥99%
Sigma-Aldrich
Tributyl phosphate, 97%
Supelco
Tributyl phosphate, Pharmaceutical Secondary Standard; Certified Reference Material
Tri-n-butyl phosphate, European Pharmacopoeia (EP) Reference Standard