MilliporeSigma
  • Energetic cost of protein import across the envelope membranes of chloroplasts.
Energetic cost of protein import across the envelope membranes of chloroplasts.
Proceedings of the National Academy of Sciences of the United States of America (2013-01-02)
Lan-Xin Shi, Steven M Theg
ABSTRACT
Chloroplasts are the organelles of green plants in which light energy is transduced into chemical energy, forming ATP and reduced carbon compounds upon which all life depends. The expenditure of this energy is one of the central issues of cellular metabolism. Chloroplasts contain ~3,000 proteins, among which less than 100 are typically encoded in the plastid genome. The rest are encoded in the nuclear genome, synthesized in the cytosol, and posttranslationally imported into the organelle in an energy-dependent process. We report here a measurement of the amount of ATP hydrolyzed to import a protein across the chloroplast envelope membranes--only the second complete accounting of the cost in Gibbs free energy of protein transport to be undertaken. Using two different precursors prepared by three distinct techniques, we show that the import of a precursor protein into chloroplasts is accompanied by the hydrolysis of ~650 ATP molecules. This translates to a ΔG(protein) (transport) of some 27,300 kJ/mol protein imported. We estimate that protein import across the plastid envelope membranes consumes ~0.6% of the total light-saturated energy output of the organelle.
Product Description
Supelco
Diuron, PESTANAL®, analytical standard
Sigma-Aldrich
Tentoxin from Alternaria tenuis, Naturally occurring phytotoxic cyclic tetrapeptide
Sigma-Aldrich
Diuron, ≥98%