MilliporeSigma
  • A randomized control trial between fosfomycin and cefuroxime as the antibiotic prophylaxis in knee arthroplasty.
A randomized control trial between fosfomycin and cefuroxime as the antibiotic prophylaxis in knee arthroplasty.
Journal of the Medical Association of Thailand = Chotmaihet thangphaet (2013-01-19)
Keerati Chareancholvanich, Pacharapol Udomkiat, Saranatra Waikakul
ABSTRACT
Antibiotic prophylaxis is used in all patient underwent total knee arthroplasty to prevent post operative infection which produced poor outcome. The suitable drug should be safe and good efficacy. To study safety and efficacy of fosfomicin and cefuroxime as antibiotic prophylaxis for total knee arthroplasty. The control trial was performed to find out efficacy and safety of fosfomycin as an antibiotic prophylaxis comparing to cefuroxime. There were 112 patients, 14 male and 98 female, with their ages ranged between 57 and 86 years. They were randomly divided into two groups, the fosfomycin group, 56 patients and the cefuroxime group, 56 patients. All patients underwent elective knee arthroplasty by the authors. The scheduled antibiotics were given perioperatively for 24 hours and all patients were followed-up for 6 months. Physical examination, skin temperature of the operated knee, radiograph and blood tests were carried out in the patients to monitor post operative infection, renal and liver disturbance. One patient in the cefuroxime group had local wound infection which responded well to local treatment and administration of antibiotics. No patients had post operative infection at the 6 months follow-up. No patients had any complication and none had renal and liver function disturbance during the follow-up. Comparing to cefuroxime,fosfomycin is safe and effective for the use as antibiotic prophylaxis in knee arthroplasty.
Product Description
Supelco
Cefuroxime, VETRANAL®, analytical standard