MilliporeSigma
  • Effects of antimineralocorticoid RU 26752 on steroid-induced hypertension in rats.
Effects of antimineralocorticoid RU 26752 on steroid-induced hypertension in rats.
The American journal of physiology (1990-05-01)
M Kalimi, J Opoku, M Agarwal, K Corley
ABSTRACT
The effect of mineralocorticoid antagonist RU 26752 on the development and maintenance of hypertension produced by long-term administration of mineralocorticoid agonist aldosterone has been investigated. Uninephrectomized, saline-drinking male Sprague-Dawley rats were subcutaneously implanted with either placebo (control) pellets or pellets containing 100 micrograms aldosterone, 50 mg RU 26752, or 100 micrograms aldosterone plus 50 mg RU 26752. Aldosterone treatment resulted in an increase in blood pressure to 165 +/- 5 mmHg over the control value of 105 +/- 2 mmHg within 3 wk of experimental period. RU 26752 given alone had no observable hypertensinogenic effect. However, RU 26752 administered with aldosterone significantly prevented the hypertension produced by aldosterone alone. RU 26752 when given with aldosterone was able to prevent the aldosterone-induced increase in saline consumption, increase urine output, and reduce urinary Na+ excretion. The results presented suggest that long-term administration of antimineralocorticoid RU 26752 in vivo to Sprague-Dawley rats prevents the aldosterone-induced hypertension.
Sigma-Aldrich
RU 26752, ≥98% (HPLC), solid